Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human SIRP gamma/CD172g Antibody, R&D Systems™
Sheep Polyclonal Antibody
Supplier: R&D Systems AF4486SP
This item is not returnable.
View return policy
Description
SIRP gamma/CD172g Polyclonal specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
SIRP gamma/CD172g | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-b2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPG, SIRPgamma, SIRP-gamma | |
Sheep | |
Affinity Purified | |
RUO | |
55423 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q9P1W8 | |
SIRPG | |
Chinese hamster ovary cell line CHO-derived recombinant human SIRP gamma/CD172g Val64-Ser364, (Val263Ala) and (Ser286Leu) Accession # Q9P1W8 | |
25 μg | |
Primary | |
Detects human SIRP gamma/CD172g in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) SIRP alpha , rhSIRP beta 1, and recombinant mouse SIRP alpha is observed. | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction