Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRPB2 Mouse anti-Human, Alexa Fluor™ 647, Clone: 1008702, R&D Systems™

Mouse Monoclonal Antibody

Manufacturer:  R&D Systems FAB99981R100UG

Catalog No. FAB99981R10


Add to cart
This item is not returnable. View return policy

Description

Description

SIRPB2 Monoclonal antibody specifically detects SIRPB2 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

SIRPB2
1008702
Flow Cytometry 0.25-1 μg/106 cells
Q5JXA9
dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG
HEK293 human embryonic kidney cell line, HEK293 derived Human SIRP beta 2, Gln33-Gly287, Accession # Q5JXA9
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human SIRP beta 2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
284759
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit