Learn More
Invitrogen™ SIRT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120542
Description
Positive test controls include: 293T, HeLa, Jurkat, NIH/3T3, C6. The target is usually found in the following locations: Cytoplasm, Mitochondrion, Nucleus, Nucleus, PML body. Immunogen sequence: VALIPSSIPH EVPQILINRE PLPHLHFDVE LLGDCDVIIN ELCHRLGGEY AKLCCNPVKL SEITEKPPRT QKELAYLSEL PPTPLHVSED SSSPERTSPP DSSVIVTLLD QAAKSNDDLD VSESKGCMEE KPQEVQTSRN VESIAEQMEN PDLKNVGSST GEKNERTSVA GTVRKCWPNR VAKEQISRRL DGNQYLFLPP NRYIFHGAEV YSDSEDDVLS SSSCGSNSDS GTCQSPSLEE PMEDESEIEE FYNGLEDEPD VPERAGGAGF GTDGDDQEAI NEAISVKQEV TDMNYPSNKS.
NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metabolism, apoptosis, and autophagy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5'-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).
Specifications
SIRT1 | |
Polyclonal | |
Unconjugated | |
SIRT1 | |
75SirT1; AA673258; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; SIR2; Sir2a; SIR2alpha; SIR2L1; sir2-like 1; SIR2-like protein 1; SIRT 1; SIRT1; SIRT-1; SirtT1 75 kDa fragment; sirtuin 1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23411 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry, Western Blot | |
2.03 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q96EB6 | |
SIRT1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 448-747 of human SIRT1 (NP_036370.2). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.