Learn More
SIRT1 Rabbit anti-Human, Unconjugated, Clone: FDA-19, Abnova™
Rabbit Monoclonal Antibody.
Manufacturer:  Abnova Corporation MAB20570
Catalog No. 89-336-970
Add to cart
Description
Description
SIRT1 Monoclonal antibody specifically detects SIRT1 in Human samples. It is validated for ICC, WB-Ce, IHC-P, IF.
Specifications
Specifications
SIRT1
FDA-19
sirtuin (silent mating type information regulation 2 homolog) 1 (S. cerevisiae)
Liquid
Rabbit
IgG
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SIRT1
SIR2L1
A synthetic phosphopeptide corresponding to residues surrounding S47 of human SIRT1
100 μg
Primary
23411