Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SIRT1 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human SIRT1 isoform 2 (Met 193-Ser 747) fused to polyhistidine tag at N-terminus

Manufacturer:  Thermo Scientific™ 11748H07E25

Catalog No. 11-748-H07E25


Add to cart

Description

Description

Expressed from DNA sequence encoding human SIRT1 isoform 2 (Met 193-Ser 747) fused to polyhistidine tag at N-terminus

  • Formulation: PBS, 10% glycerol, pH 7.4
Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
65.8kd
5 x 5μg
Full Length, Recombinant
Sirtuin 1
Assay Standard
E.coli
N-His Tag
Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
SIRT1
Other proteins
SIRT1 Recombinant Human Protein
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.