Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirt1 (Sir2) Rabbit anti-Mouse, Unconjugated, Clone: 1G16, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Sirt1 (Sir2)
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB071312MI
View Documents
MilliporeSigma
ZRB0713125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB071314MI
View Documents
MilliporeSigma
ZRB071314X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Sirt1 (Sir2) Monoclonal antibody specifically detects Sirt1 (Sir2) in Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications

Specifications

Sirt1 (Sir2)
Unconjugated
Rabbit
NAD-dependent protein deacetylase sirtuin-1;Regulatory protein SIR2 homolog 1;SIR2-like protein 1;SIR2alpha;Sir2;mSIR2a
His-tagged recombinant fragment corresponding to the first 131 amino acids from the N-terminal region of murine NAD-dependent protein deacetylase sirtuin-1 (Sirt-1).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry, Western Blot
Purified
Mouse
Sirt1, Sir2l1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit