Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT2 Rabbit anti-Human, Rat, Clone: JJ08-83, Invitrogen™

Rabbit Monoclonal Antibody

Manufacturer:  Thermo Scientific MA532451

Catalog No. PIMA532451


Add to cart

Description

Description

Rabbit Monoclonal Antibody

SIRT2 Monoclonal antibody specifically detects SIRT2 in Human, Rat samples. It is validated for Western Blot
Specifications

Specifications

SIRT2
Monoclonal
1 mg/mL
TBS with 1% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Q5RJQ4, Q8IXJ6
5730427M03Rik; mSIR2L2; NAD-dependent protein deacetylase sirtuin-2; Regulatory protein SIR2 homolog 2; SIR2; SIR2L; SIR2L2; SIR2-like protein 2; Sirt2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22933, 361532
Western Blot
JJ08-83
Unconjugated
SIRT2
Liquid
SIRT2
C terminal Human SIRT2
Protein A
RUO
Antibody
Monoclonal
Human, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit