Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT2 Rat anti-Human, Clone: 7G5, Active Motif™

Rat Monoclonal Antibody

Manufacturer:  Active MotifSupplier Diversity Partner 61366

Encompass

 View more versions of this product

Catalog No. 50-199-3349


Add to cart

Description

Description

The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.
Specifications

Specifications

SIRT2
Western Blot
1 mg/mL
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Q8IXJ6
5730427M03Rik; 5E5 antigen; mSIR2L2; NAD-dependent deacetylase sirtuin-2; NAD-dependent protein deacetylase sirtuin-2; Regulatory protein SIR2 homolog 2; silent information regulator 2; silent mating type information regulation 2, (S.cerevisiae, homolog)-like; sirtuin 2; SIR2; SIR2L; SIR2L2; SIR2-like protein 2; sir2-related protein type 2; SIRT2; sirtuin 2; sirtuin type 2; sirtuin-2
This antibody was raised against a synthetic peptide corresponding to human SIRT2 surrounding serine 331. This antibody recognizes both the unphosporylated and the Ser-331 phosphorylated form of SIRT2.
Protein G
RUO
Antibody
Monoclonal
Human
SIRT2
7G5
Unconjugated
SIRT2
Liquid
Rat
IgG2a
10 μg
-20°C, Avoid Freeze/Thaw Cycles
Primary
22933
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit