Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRT2 Rat anti-Human, Clone: 7G5, Active Motif™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

$116.85 - $356.95

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen SIRT2
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3349 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61366
10 μg
Each for $116.85
Add to cart
 
50-199-3348 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61365
100 μg
Each for $356.95
Add to cart
 
Description

Description

The SIRT2 protein (also known as Sirtuin for Silent Mating Type Information 2-Homolog) is a NAD-dependent deacytylase (NDAC) that has been shown to control gene silencing, cell cycle, and DNA damage repair. It is believed that SIRT2 may act as a tumor suppressor in human gliomas and may also serve as a novel molecular marker for these cells. SIRT2 has also been shown to act as a redox sensor to help regulate muscle gene expression in response to food intake and exercise. SIRT2 acts in the phosphorylation cascade involving mitosis where SIRT2 is phosphorylated in late G2 phase, during M phase, and into cytokinesis.
Specifications

Specifications

Monoclonal
Primary
SIRT2
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit