Learn More
SIRT6 Mouse anti-Human, Mouse, Non-human primate, Rat, Clone:, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524768
Catalog No. PIMA524768
Add to cart
Description
Description
The Silent Information Regulator (SIR2) family of genes are highly conserved from prokaryotes to eukaryotes and have important functions in the regulation of metabolism, growth and differentiation, inflammation, cellular survival, as well as in senescence and lifespan extension. Sirtuins, including SIRT1-7, are human homologs of yeast Sir2p. Sirtuins are NAD+-dependent histone/protein deacetylases (HDAC) which regulate cellular metabolism, e.g. energy metabolism, and thereby are associated with aging and several age-related diseases. SIRT6 is a histone H3 lysine 9 deacetylase and is thought to stabilize the DNA-dependent protein kinase at double-stranded DNA breaks and may have a role in the process of mammalian aging.
Specifications
Specifications
SIRT6
Monoclonal
PBS with 50% glycerol and 0.03% Proclin 300
P59941, Q8N6T7
2810449N18Rik;AI043036;Mono ADP ribosyltransferase sirtuin 6;NAD-dependent protein deacetylase sirtuin-6;Regulatory protein SIR2 homolog 6;Regulatory protein SIR2 homolog;SIR2 like 6;SIR2 like protein 6;Sir2 related protein type 6;SIR2-like protein 6;SIR2
Mouse
IgG1
100 μL
-20°C
Primary
299638, 50721, 51548
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
SIRT6
Liquid
SIRT6
Purified recombit human SIRT6 protein expressed in E.coli.
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Non-human primate, Rat