Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 2/SIRT2 Mouse anti-Human, Clone: PCRP-SIRT2-1A8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30735810UG

 View more versions of this product

Catalog No. NB122189


Add to cart

Description

Description

Sirtuin 2/SIRT2 Monoclonal antibody specifically detects Sirtuin 2/SIRT2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

Sirtuin 2/SIRT2
PCRP-SIRT2-1A8
Unconjugated
10 mM PBS with 0.05% BSA
EC 3.5.1, EC 3.5.1.-, FLJ35621, FLJ37491, S.cerevisiae, homolog) 2, sirtuin 2, sirtuin type 2
Recombinant full-length human Sirtuin 2/SIRT2 protein
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG1
100 μg
Apoptosis, Cancer, Chromatin Research, DNA Repair, Epigenetics, Neuronal Cell Markers, Transcription Factors and Regulators
Primary
22933
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit