Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sirtuin 2/SIRT2, Mouse anti-Human, Clone: S2R233-1, Novus Biologicals™

Mouse Monoclonal Antibody

$240.50 - $314.00

Specifications

Antigen Sirtuin 2/SIRT2
Clone S2R233-1
Applications Western Blot, ELISA
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013174
View Documents
Novus Biologicals
NBP280049100UG
100 ug
Each for $314.00
Add to cart
 
NB013175
View Documents
Novus Biologicals
NBP28004950UG
50 ug
Each for $240.50
Add to cart
 
Description

Description

Sirtuin 2/SIRT2 Monoclonal antibody specifically detects Sirtuin 2/SIRT2 in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Sirtuin 2/SIRT2
Western Blot, ELISA
Human
Primary
S2R233-1
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit