Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sitagliptin Phosphate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Click to view available options
Quantity:
1 g
Specifications
Specifications
CAS | 654671-77-9 |
Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
Linear Formula | C16H15F6N5 O · H3 PO4 · H2O |
Synonym | Sitagliptin phosphate monohydrate; 7-[(3 R)-3-amino-1-oxo-4-(2,4,5-trifluorophenyl)butyl]-5,6,7,8-tetrahydro-3-(trifluoromethyl)-1,2,4-triazolo[4,3-a]pyrazine phosphate; MK-0431 |
Type | Certified Reference Material |
Quantity | 1 g |
Formula Weight | 523.32 |
Grade | Certified Reference Material |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction