Learn More
Apexbio Technology LLC Sitaxentan sodium(Synonyms: TBC11251, Sitaxsentan sodium, Thelin, Sitaxentan sodium hydrate, Sitaxentan sodium salt), 10mg, CAS: 210421-74-2.
SDP
Supplier:  Apexbio Technology LLC B152210
Encompass_Preferred
Sitaxentan sodium (CAS 210421-74-2) is a small molecule compound functioning as an orally bioavailable and highly selective antagonist for endothelin receptor subtype A (ETA) By specifically inhibiting ETA receptor signaling sitaxentan sodium interferes with endothelin-1 mediated vasoconstriction and cellular proliferation pathways In biomedical research contexts it serves as a valuable tool for investigating endothelin-related physiological and pathological processes with particular relevance to studies on vascular disorders and pulmonary arterial hypertension
Catalog No. 50-000-60562
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.