Learn More
Fisherbrand™ Sitting Height Wood Fixed Height Table, 60 in. Wide, Black Epoxy Work Surface
Fisherbrand™ Wood Fixed Height Tables bring quality, warmth and durability to your laboratory.
Description
- Made from hand selected premium rotary cut maple or rift cut oak veneers.
- Robotically applied environmentally friendly finishes are chemically resistant.
- Wood Fixed Height Tables are furnished with 4.875 in. tall apron rails.
- The legs have a 2.25 x 2.25 in. construction and inculdes leveling feet and a black trim boot at the base of the leg.
Compliance
SEFA 8
Specifications
Fixed Height Table | |
Honey Stain | |
30 in. | |
30 in. | |
60 in. | |
Oak Veneer |
Table Frame, Epoxy Work Surface, Leveling Feet and Black Trim Boot | |
76.2 cm | |
76.2 cm | |
2 years | |
154.9 cm |