Learn More
Fisherbrand™ Sitting Height Wood Knee Space Panels for use with Drawer Apron Frames, 36 in. Wide
Fisherbrand™ Wood Knee Space Panels bring quality, warmth and durability to your laboratory.
 View more versions of this product
Catalog No. FB902436M6
Add to cart
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Knee space panels close off the casework to the call to create a clear sitting or standing space under the work surface. 
Compliance
SEFA 8
Specifications
Specifications
Chili Stain
2.5 cm
23.75 in.
Wood (Maple Veneer)
91.4 cm
1 in.
Drawer Apron Frames
60.3 cm
36 in