Learn More
BioworldSupplier Diversity Partner SIV P55 (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220606971
Recombinant SIV p55 Source: Insect Cells Description : Recombinant SIVp55, Strains: SIV mac 23g and SIV smH4 is glycosylated with N-linked sugars and produced using baculovirus vectors in insect cells. SIV-1 p66 is purified by propri...
Catalog No. 50-253-798