Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIVA1 Monoclonal Antibody (OTI1B10), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526278
Description
SIVA1 Monoclonal antibody specifically detects SIVA1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
SIVA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15304 | |
SIVA1 | |
Full length human recombinant protein of SIVA1 produced in HEK293T cell | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B10 | |
Unconjugated | |
SIVA1 | |
apoptosis regulatory protein Siva; Cd27 binding protein; CD27-binding (Siva) protein; CD27-binding protein; CD27BP; SIVA; SIVA1; Siva-1; SIVA1 apoptosis inducing factor; SIVA1, apoptosis-inducing factor; Siva-2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10572 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction