Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIVA1 Mouse anti-Human, Clone: OTI1C9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526277

Catalog No. PIMA526277


Add to cart

Description

Description

Induces CD27-mediated apoptosis. Inhibits BCL2L1 isoform Bcl-x(L) anti-apoptotic activity. Inhibits activation of NF-kappa-B and promotes T-cell receptor-mediated apoptosis.
Specifications

Specifications

SIVA1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O15304
SIVA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10572
Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C9
Unconjugated
SIVA1
Liquid
SIVA1
Full length human recombit protein of SIVA1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit