Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIX1 Monoclonal Antibody (OTI1H4)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525811

Catalog No. PIMA525811


Only null left
Add to Cart

Description

Description

SIX1 Monoclonal Antibody for Western Blot

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal dominant deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIX1
Monoclonal
0.77 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q15475
SIX1
Human recombinant protein fragment corresponding to amino acids 1-284 of SIX1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI1H4
Unconjugated
SIX1
BB138287; BOS3; DFNA23; Homeobox protein SIX1; sine oculis homeobox homolog 1; sine oculis-related homeobox 1; sine oculis-related homeobox 1 homolog; SIX homeobox 1; SIX1; TIP39
Mouse
Affinity Chromatography
RUO
6495
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.