Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX1 Mouse anti-Human, Clone: OTI1H4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504057S

Catalog No. 50-167-4828


Add to cart

Description

Description

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila  ft.sine oculis ft. gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal domit deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3).
Specifications

Specifications

SIX1
Monoclonal
0.77 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q15475
BOS3, DFNA23, TIP39
Mouse
IgG1
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6495
Western Blot
OTI1H4
Unconjugated
SIX1
Liquid
SIX1
Human recombit protein fragment corresponding to amino acids 1-284 of human SIX1 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit