Learn More
Invitrogen™ SIX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595466
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a member of the sine oculis homeobox transcription factor family. The encoded protein plays a role in eye development. Mutations in this gene have been associated with holoprosencephaly type 2.
Specifications
SIX3 | |
Polyclonal | |
Unconjugated | |
Six3 | |
E130112M24Rik; Homeobox protein SIX3; HPE2; sine oculis homeobox 3-like protein; sine oculis homeobox homolog 3; sine oculis homeobox-like protein 3; sine oculis-related homeobox 3; sine oculis-related homeobox 3 homolog; SIX homeobox 3; Six3; Six3a; Six3alpha; Six3b; Six3beta | |
Rabbit | |
Affinity chromatography | |
RUO | |
20473, 6496, 78974 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and 0.05mg sodium azide | |
O95343, Q62233 | |
Six3 | |
A synthetic peptide corresponding to a sequence at the N-terminus of human Six3 (1-32aa MVFRSPLDLYSSHFLLPNFADSHHRSILLASS). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.