Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIX4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540538
Description
Peptide sequence: EKSRNALKEL YKQNRYPSPA EKRHLAKITG LSLTQVSNWF KNRRQRDRNP Sequence homology: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.
SIX4 gene ontology annotations related to this gene include anatomical structure development; embryonic cranial skeleton morphogenesis; fungiform papilla morphogenesis; generation of neurons; inner ear morphogenesis; male gonad development; male sex determination; metanephric mesenchyme development; myoblast migration; myotome development; negative regulation of neuron apoptotic process; negative regulation of satellite cell differentiation; negative regulation of transcription, DNA-templated; olfactory placode formation; pharyngeal system development; positive regulation of branching involved in ureteric bud morphogenesis; positive regulation of ureteric bud formation; protein localization to nucleus; regulation of branch elongation involved in ureteric bud branching; regulation of epithelial cell proliferation; regulation of protein localization; regulation of synaptic growth at neuromuscular junction; sarcomere organization; skeletal muscle fiber differentiation; thymus development; trigeminal ganglion development.
Specifications
SIX4 | |
Polyclonal | |
Unconjugated | |
SIX4 | |
AI047561; Arec3; homeobox protein SIX4; Sine oculis homeobox homolog 4; sine oculis-related homeobox 4; sine oculis-related homeobox 4 homolog; SIX homeobox 4; Six4; Skeletal muscle-specific ARE-binding protein AREC3; TrexBF | |
Rabbit | |
Affinity Chromatography | |
RUO | |
51804 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UIU6 | |
SIX4 | |
Synthetic peptide directed towards the middle region of human SIX4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction