Learn More
Invitrogen™ ACTA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114172
Description
Immunogen sequence: GQKDSYVGDE AQSKRGILTL KYPIEHGIIT NWDDMEKIWH HTFYNELRVA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Rabbit: 100%; Rat: 100%.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
Specifications
Skeletal Muscle Actin | |
Polyclonal | |
Unconjugated | |
Acta1 | |
0610041G09Rik; AA959943; AAT6; ACTA; Acta1; ACTA1 protein; Acta2; Acta-2; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; Acts; ACTSA; Actsk-1; Actvs; alpha actin; alpha actin 2; Alpha-actin-1; Alpha-actin-2; alpha-cardiac actin; alphaSMA; alpha-smooth muscle actin; ASMA; a-SMA; cell growth-inhibiting gene 46 protein; CFTD; CFTD1; CFTDM; cytoplasmic beta-actin; GIG46; hypothetical protein LOC515610; LOW QUALITY PROTEIN: actin, alpha skeletal muscle; LOW QUALITY PROTEIN: actin, aortic smooth muscle; MPFD; MYMY5; NEM1; NEM2; NEM3; nemaline myopathy type 3; SHPM; SMalphaA; smooth muscle alpha-actin; unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
11459, 11475, 58, 59 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P62736, P62737, P68133, P68134 | |
Acta1, ACTA2 | |
Immunogen sequence: GQKDSYVGDE AQSKRGILTL KYPIEHGIIT NWDDMEKIWH HTFYNELRVA (aa 50-99). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.