Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKF 77434 hydrobromide, Tocris Bioscience™

Catalog Number 166210
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
300561-58-4
C19H22BrNO2
376.294
JWQRAXTWDYUBFI-UHFFFAOYSA-N
skf 77434 hydrobromide, n-allyl-+/--skf-38393 hydrobromide, skf-77434 hydrobromide, n-allyl-inverted exclamation marka-skf-38393 hydrobromide, 3-allyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, n-allyl-1-phenyl-2,3,4,5-tetrahydro-3-benzazepine-7,8-diol hydrobromide, 1-phenyl-3-prop-2-en-1-yl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 3-allyl-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide, n-allyl-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrobromide, +/--skf 38393, n-allyl-, hydrobromide
11957703
CHEBI:63978
5-phenyl-3-prop-2-enyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide
C=CCN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)O.Br
This item is not returnable. View return policy
300561-58-4
C19H22BrNO2
376.294
JWQRAXTWDYUBFI-UHFFFAOYSA-N
skf 77434 hydrobromide, n-allyl-+/--skf-38393 hydrobromide, skf-77434 hydrobromide, n-allyl-inverted exclamation marka-skf-38393 hydrobromide, 3-allyl-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, n-allyl-1-phenyl-2,3,4,5-tetrahydro-3-benzazepine-7,8-diol hydrobromide, 1-phenyl-3-prop-2-en-1-yl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 3-allyl-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide, n-allyl-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrobromide, +/--skf 38393, n-allyl-, hydrobromide
11957703
CHEBI:63978
5-phenyl-3-prop-2-enyl-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide
C=CCN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)O.Br

Selective D1-like partial agonist

Selective dopamine D1-like receptor partial agonist (IC50 values are 19.7 and 2425 nM for binding to D1-like and D2-like receptors respectively). Centrally active following systemic administration in vivo.
Quantity 10 mg
Formula Weight 376.29
Percent Purity >99%
Chemical Name or Material SKF 77434 hydrobromide
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit