Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SKF 82958 hydrobromide Available on GSA/VA Contract for Federal Government customers only.

D1 agonist

Manufacturer:  Tocris Bioscience™ 5719/50

 View more versions of this product

Catalog No. 57-195-0


Add to cart
This item is not returnable. View return policy

Description

Description

D1 agonist. Increases renal blood flow in animal models and promotes contralateral rotation in rats with unilateral lesions of the substantia nigra. Also augments GABAB inhibitory post-synaptic potentials (ipsp) in ventral tegmental area in brain slices.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

6-Chloro-2,3,4,5-tetrahydro-1-phenyl-3-(2-propen-1-yl)-1H-3-benzazepine-7,8-diol hydrobromide
C19H20ClNO2.HBr
50mg
>98%
74115-01-8
410.73
Target: Dopamine D1 and D5 Receptor Antagonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at RT

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit