Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKF 86002 dihydrochloride, Tocris Bioscience™

Catalog Number 200810
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
116339-68-5
C16H14Cl2FN3S
370.267
GQQCNUNCYVXBTF-UHFFFAOYSA-N
skf 86002 dihydrochloride, 6-4-fluorophenyl-2,3-dihydro-5-4-pyridinyl imidazo 2,1-b thiazole dihydrochloride, 6-4-fluorophenyl-5-pyridin-4-yl-2,3-dihydroimidazo 2,1-b thiazole dihydrochloride
10339107
6-(4-fluorophenyl)-5-pyridin-4-yl-2,3-dihydroimidazo[2,1-b][1,3]thiazole;dihydrochloride
C1CSC2=NC(=C(N21)C3=CC=NC=C3)C4=CC=C(C=C4)F.Cl.Cl
This item is not returnable. View return policy
116339-68-5
C16H14Cl2FN3S
370.267
GQQCNUNCYVXBTF-UHFFFAOYSA-N
skf 86002 dihydrochloride, 6-4-fluorophenyl-2,3-dihydro-5-4-pyridinyl imidazo 2,1-b thiazole dihydrochloride, 6-4-fluorophenyl-5-pyridin-4-yl-2,3-dihydroimidazo 2,1-b thiazole dihydrochloride
10339107
6-(4-fluorophenyl)-5-pyridin-4-yl-2,3-dihydroimidazo[2,1-b][1,3]thiazole;dihydrochloride
C1CSC2=NC(=C(N21)C3=CC=NC=C3)C4=CC=C(C=C4)F.Cl.Cl

p38 MAPK inhibitor; anti-inflammatory agent

Inhibitor of p38 MAP kinase (IC50 = 0.1 - 1 μM). Potently inhibits LPS-induced IL-1 and TNF-α production in human monocytes (IC50 = 1 μM). Also inhibits 5-lipoxygenase- and cyclooxygenase-mediated arachidonic acid metabolism in RBL-1 cells (IC50 values are 10 and 100 μM respectively). Anti-inflammatory following oral administration in vivo.
Quantity 10 mg
Formula Weight 370.27
Percent Purity >98%
Chemical Name or Material SKF 86002 dihydrochloride
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit