Learn More
Tocris Bioscience™ SKF 91488 dihydrochloride
Histamine N-methyltransferase inhibitor
Supplier: Tocris Bioscience™ 0512/10
This item is not returnable.
View return policy
Description
SKF 91488 dihydrochloride is a homolog of dimaprit (Cat. No. 0506); strong inhibitor of histamine N-methyltransferase. Exhibits no histamine agonist activity.Specifications
4-(N,N-Dimethylamino)butylisothiourea dihydrochloride | |
10 mg | |
C7H17N3S.2HCl |
68941-21-9 | |
Soluble to 75mM in DMSO | |
Observed MW: 248.21 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature