Learn More
Tocris Bioscience™ SKF 91488 dihydrochloride
Histamine N-methyltransferase inhibitor
$160.00 - $574.00
Specifications
Chemical Name or Material | 4-(N,N-Dimethylamino)butylisothiourea dihydrochloride |
---|---|
CAS | 68941-21-9 |
Solubility | Soluble to 75mM in DMSO |
Molecular Formula | C7H17N3S.2HCl |
Formula Weight | Observed MW: 248.21 |
Description
Homolog of dimaprit (Cat. No. 0506); strong inhibitor of histamine N-methyltransferase. Exhibits no histamine agonist activity.Specifications
4-(N,N-Dimethylamino)butylisothiourea dihydrochloride | |
Soluble to 75mM in DMSO | |
Observed MW: 248.21 |
68941-21-9 | |
C7H17N3S.2HCl |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.