Learn More
Apexbio Technology LLC SKF38393 HCl,100mg CAS# 62717-42-4
SDP
Supplier:  Apexbio Technology LLC B6030100
Encompass_Preferred
SKF38393 HCl (SKF38393) is a selective dopamine D1 receptor agonist [1], with an IC50 value of 110 nM [2].There are two functional types of dopamine receptors, D1 and D2 receptors [3]. The stimulation of the D1 receptor (D1R) in prefrontal cortex (PFC) results in an ‘inverted-U’ dose-response. Either too little or too much D1R stimulation can impair spatial working memory [4].6-OHDA reduced the specific expression of mRNAs to encode substance P and D1 dopamine receptor in striatonigral neurons. Subsequently daily injections of SKF-38393 reversed this reduction effect. The treatment with SKF-38393 also increased dynorphin mRNA [5]. D1Rs activate cyclic AMP-dependent protein kinases, stimulate adenylyl cyclase, regulate neuron growth and differentiation, modify dopamine D2 receptor-mediated events and influence behavior [3]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0504
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.