Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR61 Rabbit anti-Drosophila, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA540079

Catalog No. PA540079


Add to cart

Description

Description

SKI8 is a component of the PAF and SKI complex which is composed of SKI2, SKI3 and SKI8. These are involved in transcriptional regulation and events downstream of RNA synthesis. Interaction of the SKI complex with SKI7 links the SKI complex to the exosome in order to perform mRNA degradation.
Specifications

Specifications

WDR61
Polyclonal
Unconjugated
WDR61
Liquid
WDR61
Recombit human SKI8 protein.
Protein G
RUO
Antibody
Polyclonal
Drosophila, Human
ChIP assay, Western Blot
2.5 mg/mL
PBS with 0.05% Proclin 300, 0.05% sodium azide; pH 7.4
Q9GZS3
WDR61, REC14WD repeat-containing protein 61, Meiotic recombination REC14 protein homolog, SKI8 homolog
Rabbit
IgG
150 μg
-20°C or -80°C if preferred
Primary
80349
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit