SHOWA™ Skinny Dip™ Gloves
Outstanding dexterity with greater comfort
Manufacturer: SHOWA™ 381110
 View more versions of this product
Catalog No. 19-050-149
  Check Availability
Add to Cart
    Specifications
    Cleaning Procedure Launder at 40°C using a neutral Detergent
    Exterior Finish Wrinkle
    Glove Cuff Style Knit
    For Use With (Application) construction, carpentry, maintenance, highway work, landscaping, floor tiling, garbage collection, roofing, shipping and receiving
    Material Natural Rubber
    Liner Material Cotton
    Size 10
    Coated Area Palm
    Color Gray
    Cut Resistance Rating Level 2
  • Description & Specifications
Related Products
Product Content Feedback