Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKIP Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174128
Description
SKIP Polyclonal specifically detects SKIP in Human samples. It is validated for Western Blot.Specifications
SKIP | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9BT40 | |
INPP5K | |
Synthetic peptides corresponding to the C terminal of SKIP. Immunizing peptide sequence PPTYKFDRNSNDYDTSEKKRKPAWTDRILWRLKRQPCAGPDTPIPPASHF. | |
Affinity Purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Rabbit: 86%; Horse: 85%; Guinea pig: 85%; Yeast: 83%. | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
EC 3.1.3, EC 3.1.3.56, inositol polyphosphate-5-phosphatase K, PPS, skeletal muscle and kidney enriched inositol phosphatase, Skeletal muscle and kidney-enriched inositol phosphatase, SKIPinositol polyphosphate 5-phosphatase K | |
Rabbit | |
49 kDa | |
100 μL | |
Protein Phosphatase | |
51763 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title