Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKIV2L Mouse anti-Human, Clone: 1E5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524551

Catalog No. PIMA524551


Add to cart

Description

Description

Peptide Sequence: DQLPNTLKQG IERVRAVAKR IGEVQVACGL NQTVEEFVGE LNFGLVEVVY EWARGMPFSE LAGLSGTPEG LVVRCIQRLA EMCRSLRGAA RLVGEPVLGA KMETAATLL

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a human homologue of yeast SKI2 and may be involved in antiviral activity by blocking translation of poly(A) deficient mRNAs. This gene is located in the class III region of the major histocompatibility complex.
Specifications

Specifications

SKIV2L
Monoclonal
Unconjugated
SKIV2L
Liquid
SKIV2L
SKIV2L (NP_008860, 1125 a.a. ∽ 1233 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
200 μL
-20° C
Primary
6499
ELISA, Western Blot
1E5
ascites with no preservative
Q15477
170A, DDX13, HLP, SKI2, SKI2W, SKIV2, helicase SKI2W, helicase-like protein
Mouse
IgG2a, kappa
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit