Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SKIV2L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541280
Description
Peptide sequence: KGPPGSADKA GKRFDGKLQS ESTNNGKNKR DVDFEGTDEP IFGKKPRIEE Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 85%; Rabbit: 100%.
SKIV2L2 may be involved in pre-mRNA splicing.
Specifications
SKIV2L2 | |
Polyclonal | |
Unconjugated | |
MTREX | |
2610528A15Rik; ATP-dependent helicase SKIV2L2; ATP-dependent RNA helicase DOB1; ATP-dependent RNA helicase SKIV2L2; Dob1; Exosome RNA helicase MTR4; exosome RNA helicase MTR4; superkiller viralicidic activity 2-like 2; fSAP118; functional spliceosome-associated protein 118; KIAA0052; mKIAA0052; Mtr4; Mtr4 exosome RNA helicase; MTREX; RGD1305984; Ski2 like RNA helicase 2; SKIV2L2; Superkiller viralicidic activity 2-like 2; superkiller viralicidic activity 2-like 2 (S. cerevisiae); superkiller viralicidic activity 2-like 2-like; TRAMP-like complex helicase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23517 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P42285 | |
MTREX | |
Synthetic peptide directed towards the N-terminal of human SKIV2L2 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction