Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SKIV2L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549633

Catalog No. PA549633

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

SKIV2L2 may be involved in pre-mRNA splicing.
Specifications

Specifications

SKIV2L2
Polyclonal
PBS with 0.09% sodium azide
P42285
Superkiller viralicidic activity 2-like 2, ATP-dependent RNA helicase SKIV2L2, TRAMP-like complex helicase, SKIV2L2, KIAA0052, Mtr4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23517
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SKIV2L2
Liquid
SKIV2L2
KLH conjugated synthetic peptide between 237-263 amino acids from the Central region of human SKIV2L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit