Learn More
SKP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150101AP
Catalog No. 50-172-9907
Add to cart
Description
Description
Skp2 (S-phase Kinase-associated Protein 2) belongs to the family of F-box proteins that interact with the Cyclin A-Cdk2 complex. Skp2 is essential for the G1-S transition in both transformed cells and diploid fibroblasts. Biochemical and genetic experiments have demonstrated that Skp2 is required for the ubiquitination and consequent degradation of p27 in cultured mammalian cells and in vitro reconstitution assays. In normal tissues, Skp2 is expressed in tonsil and placenta. In tumor tissues, Skp2 is expressed widely, in some colon, prostate, pancreas, and skin cancers, with high expression in lung, breast, ovarian, and endometrial cancers. Research suggests that Skp2 plays a role in oncogenesis, particularly in the pathogenesis of lymphomas and breast cancers.
Specifications
Specifications
SKP2
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13309, Q9Z0Z3
F box protein Skp2, F box/LRR repeat protein 1, FBL1, FBXL1, FLB1, p45skp2, SKP2
Rabbit
IgG
150 μL
-20°C
Primary
27401, 294790, 6502
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
SKP2
Liquid
Skp2
SKP2 Fusion Protein Ag6826
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat