Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SKPin C1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of Skp2-mediated p27 degradation. Induces p27 accumulation in metastatic melanoma cell lines. Promotes G1/S cell cycle arrest in T47D cells and LNCaP cells; induces G2/M cell cycle arrest in MCF-7 cells.
Specifications
Quantity | 10 mg |
Formula Weight | 465.34 |
Percent Purity | >98% |
Chemical Name or Material | SKPin C1 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction