Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLAIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566605
Description
Immunogen sequence: SPARGIEYSRV SPQPMISRLQ QPRLSLQGHP TDLQTSNVKN EEKLRRSLPN LSRTSNTQVD SVKSSRSDSN FQVPN Highest antigen sequence identity to the following orthologs - mouse 97%, rat 99%.
SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
Specifications
SLAIN2 | |
Polyclonal | |
Unconjugated | |
SLAIN2 | |
5033405K12Rik; 8030444K12Rik; AI596370; FLJ21611; hypothetical protein LOC534203; KIAA1458; LOW QUALITY PROTEIN: SLAIN motif-containing protein 2; RGD1311593; SLAIN motif family member 2; SLAIN motif family, member 2; SLAIN motif-containing protein 2; SLAIN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57606 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P270 | |
SLAIN2 | |
Recombinant Human SLAIN2. Recombinant protein control fragment (Product #RP-107260). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction