Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals SLAM/CD150 Antibody Pair [HRP]
Monoclonal Antibody Pair
Manufacturer: Novus Biologicals NBP2793075PLATES
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SLAM/CD150.Specifications
7.8125 pg/mL | |
Sandwich ELISA | |
RUO | |
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human SLAM/CD150. | |
SLAM/CD150 | |
Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
Human | |
SLAMF1 | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 0.5 ng/mL is recommended. |
HRP | |
6504 | |
Antibody Pairs | |
7.81-500 pg/mL | |
Storage is content dependent. | |
Mouse/Rabbit | |
5 Plates | |
Sandwich ELISA | |
CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title