Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAM/CD150 Mouse anti-Human, Janelia Fluor 549, Clone: SLAM.4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262198JF549
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
SLAM/CD150 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLAMF1 | |
Human CD150-transfected 300.19 cells | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry | |
SLAM.4 | |
Flow Cytometry, Immunocytochemistry | |
CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
Mouse | |
Protein A purified | |
Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Stem Cell Markers | |
6504 | |
Store at 4°C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title