Learn More
Eisco™ Slate Specimen
SCI_ED
Slate Specimen for use in metamorphic classification and identification exercises.
Supplier:  Eisco™ ESNG0063PK10
 View more versions of this product
Catalog No. S26601
Add to Cart
Description
Description
  • For use in metamorphic rock identification exercises due to slate's recognizable platy foliation
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Differentiating between slate and shale offers interesting exercise in determining rock physical properties
  • Forms from compression of shale
  • Denser than shale
  • Retains the fissile properties of shale
Specifications
Specifications
Metamorphic Rock, Slate
10/Pk.
2 to 3 cm
0.2 kg