Learn More
Eisco™ Slate Specimen
Slate Specimen for use in metamorphic classification and identification exercises.
$2.85 - $30.75
Catalog Number | Mfr. No. | Specimen Size | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Specimen Size | Quantity | Price | Quantity & Availability | ||||
S24796
![]() |
Eisco™
ESNG0063 |
2 to 3 cm | 1 Ea. |
Each for $3.00
|
|
||||
S26601
![]() |
Eisco™
ESNG0063PK10 |
2 to 3 cm | 10/Pk. |
Each for $2.85
|
|
||||
S24797
![]() |
Eisco™
ESNG0063PK12 |
2 to 3 cm | 12/Pk. |
Pack of 12 for $30.75
|
|
||||
S26624
![]() |
Eisco™
ESNG0063HAND |
7 to 8 cm | 1 Ea. |
Each for $16.50
|
|
||||
Description
- For use in metamorphic rock identification exercises due to slate's recognizable platy foliation
- For use in classroom identification labs and exercises
- Correlates well to Next Generation Science Standards Evidence Statements
- Differentiating between slate and shale offers interesting exercise in determining rock physical properties
- Forms from compression of shale
- Denser than shale
- Retains the fissile properties of shale
Specifications
Metamorphic Rock, Slate |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.