Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC10A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100125
Description
Antibody detects endogenous levels of total ASBT.
This gene encodes a sodium/bile acid cotransporter. This transporter is the primary mechanism for uptake of intestinal bile acids by apical cells in the distal ileum. Bile acids are the catabolic product of cholesterol metabolism, so this protein is also critical for cholesterol homeostasis. Mutations in this gene cause primary bile acid malabsorption (PBAM); mutations in this gene may also be associated with other diseases of the liver and intestines, such as familial hypertriglyceridemia (FHTG).
Specifications
SLC10A2 | |
Polyclonal | |
Unconjugated | |
Slc10a2 | |
Apical sodium-dependent bile acid transporter; ASBT; IBAT; ileal apical sodium-dependent bile acid transporter; ileal Na(+)/bile acid cotransporter; ileal sodium/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; ISBAT; ISBT; Na(+)-dependent ileal bile acid transporter; NTCP2; PBAM; Slc10a2; Sodium/taurocholate cotransporting polypeptide, ileal; sodium/taurocholate-cotransporting polypeptide, ileal; solute carrier family 10 (sodium/bile acid cotransporter family), member 2; solute carrier family 10 (sodium/bile acid cotransporter), member 2; solute carrier family 10 member 2; solute carrier family 10, member 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
20494, 6555 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P70172, Q12908 | |
Slc10a2 | |
A synthesized peptide derived from human SLC10A2(Accession Q12908), corresponding to amino acid residues P332-K348. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction