Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC10A5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | SLC10A5 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15970720
|
Novus Biologicals
NBP15970720UL |
20 μL |
Each for $204.00
|
|
|||||
NBP159707
|
Novus Biologicals
NBP159707 |
100 μL |
Each for $482.50
|
|
|||||
Description
SLC10A5 Polyclonal specifically detects SLC10A5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SLC10A5 | |
Polyclonal | |
Purified | |
RUO | |
Na(+)/bile acid cotransporter 5, P5, sodium/bile acid cotransporter 5, solute carrier family 10 (sodium/bile acid cotransporter family), member 5, Solute carrier family 10 member 5 | |
SLC10A5 | |
Primary | |
48 kDa |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Q5PT55 | |
347051 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title