Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC12A8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$157.00 - $482.50
Specifications
Antigen | SLC12A8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15989320
|
Novus Biologicals
NBP15989320UL |
20 μL |
Each for $157.00
|
|
|||||
NBP159893
|
Novus Biologicals
NBP159893 |
100 μL |
Each for $482.50
|
|
|||||
Description
SLC12A8 Polyclonal specifically detects SLC12A8 in Human samples. It is validated for Western Blot.Specifications
SLC12A8 | |
Polyclonal | |
Rabbit | |
A0AV02 | |
84561 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
CCC9member 8, DKFZp686L18248, FLJ23188, solute carrier family 12 (potassium/chloride transporters), member 8, solute carrier family 12 member 8 | |
SLC12A8 | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title