Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC16A13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566755
Description
Immunogen sequence: PPSLAEDPAVG GPRAQLTSLL HHGPF Highest antigen sequence identity to the following orthologs - mouse 77%, rat 77%.
Monocarboxylates, such as lactate and pyruvate, play an integral role in cellular metabolism. Lactic acid is produced in large quantities as a result of glycolysis, which provides the majority of ATP to cells under normal physiological conditions. However, accumulation of lactic acid leads to a decrease in intracellular pH and, thus, to a cessation of glycolysis. In order for glycolysis to continue at a high rate, lactic acid must be transported out of the cell. This transport process is carried out by a family of monocarboxylate transporters (MCTs), which function as proton symports and are stereoselective for L-lactate. MCT13 (Monocarboxylate transporter 13), also known as SLC16A13 (Solute carrier family 16 member 13), is a 426 amino acid multi-pass membrane protein that belongs to the MCT transport family. Functioning as a proton-linked monocarboxylate transporter, MCT13 catalyzes the rapid transports of molecules, such as lactate, across the plasma membrane.
Specifications
SLC16A13 | |
Polyclonal | |
Unconjugated | |
SLC16A13 | |
1700007D07Rik; MCT 13; MCT13; monocarboxylate transporter; Monocarboxylate transporter 13; monocarboxylic acid transporter 13; MOT13; Slc16a13; solute carrier family 16 (monocarboxylic acid transporters), member 13; solute carrier family 16 member 13; solute carrier family 16, member 13; solute carrier family 16, member 13 (monocarboxylic acid transporter 13) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
201232 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7RTY0 | |
SLC16A13 | |
Recombinant Human SLC16A13. Recombinant protein control fragment (Product #RP-107413). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction