Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC16A4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550519

Catalog No. PA550519


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC16A4 protein.

Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate
Specifications

Specifications

SLC16A4
Polyclonal
Unconjugated
SLC16A4
Liquid
SLC16A4
Synthetic peptide corresponding to residues near the N terminal of human solute carrier family 16, member 4 (monocarboxylic acid transporter 5)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
O15374
MCT4, MCT5
Rabbit
IgG
100 μL
-20°C
Primary
9122
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit