Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC17A1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550829

Catalog No. PA550829


Add to cart

Description

Description

The antibody detects endogenous levels of total SLC17A1 protein.

The function of this protein remains unknown.
Specifications

Specifications

SLC17A1
Polyclonal
Unconjugated
SLC17A1
Liquid
SLC17A1
Synthetic peptide corresponding to residues near the C terminal of human solute carrier family 17 (organic anion transporter), member 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
2.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q14916
NPT1, NPT-1, NAPI-1
Rabbit
IgG
100 μL
-20°C
Primary
6568
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit