Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC17A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143477
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
SLC17A3 may be involved in actively transporting phosphate into cells via Na(+) cotransport.
Specifications
SLC17A3 | |
Polyclonal | |
Unconjugated | |
SLC17A3 | |
GOUT4; Na(+)/PI cotransporter 4; NPT4; SLC17A3; sodium/phosphate cotransporter 4; Sodium-dependent phosphate transport protein 4; solute carrier family 17 (organic anion transporter), member 3; solute carrier family 17 (sodium phosphate), member 3; solute carrier family 17 member 3; UAQTL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105355, 10786, 266730 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.92 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
O00476 | |
SLC17A3 | |
Recombinant protein encompassing a sequence within the center region of human SLC17A3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction